Learn More
Fisherbrand™ accumet™ AE150 Benchtop pH Meter
Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price
357.00 € - 465.00 €
Specifications
Accuracy (mV) | ±0.2 ±199.9 ±2 beyond ±200 |
---|---|
Outputs | Recorder (±2000 mV) |
Range (pH) | 0 to 14 |
Range (mV) | ±1999 |
Resolution (mV) | 0.1 mV (±199.9 mV); 1 mV beyond ±200 mV |
Product Code | Brand | Includes | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Type | Price | Quantity & Availability | ||||||
15514693
|
Fisherbrand™
13-636-AE150 |
Meter, Electrode Stand, Univ. Power Adapter (90-240V, 50/60 Hz) | Standard Set | N/A | N/A | N/A | |||||
15524693
|
Fisherbrand™
13-636-AE151 |
Meter, temperature probe and stand | Education Set | N/A | N/A | N/A | |||||
15534693
|
Fisherbrand™
13-636-AE152 |
Meter, Double Junction pH Electrode, Temperature Probe and Stand | Bio Set | N/A | N/A | N/A | |||||
15544693
|
Fisherbrand™
13-636-AE153 |
Meter, 13-620-AE6 3-in-1 single-junction gel pH/ATC electrode, and stand | 3-in-1 Set | N/A | N/A | N/A | |||||
Description
Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet AE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.
- One to five point pH calibration
- Automatic buffer recognition with two pH buffer sets
- pH slope and offset display
- One point mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation
- Ready icon to indicate measurement stability
- Electrode condition icon to indicate status
- 100 data set storage with non-volatile memory
Warranty
3 years on meter
Specifications
±0.2 ±199.9 ±2 beyond ±200 | |
0 to 14 | |
0.1 mV (±199.9 mV); 1 mV beyond ±200 mV | |
0.1°C | |
100 data sets | |
AC/DC 9 V adapter (110/220 VAC, 50-60 Hz) |
Recorder (±2000 mV) | |
±1999 | |
0.0°C to 100.0°C | |
Offset in 0.1 °C increments; Offset range: ±5 °C | |
BNC, 2.5 mm phono plug, reference pin tip, ground |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.