Learn More
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals
34.87 € - 274.37 €
Chemical Identifiers
CAS | 58052-80-5 |
---|---|
Molecular Formula | C6H18N3 |
Molecular Weight (g/mol) | 132.23 |
MDL Number | MFCD00149559 |
InChI Key | MZSSRMMSFLVKPK-UHFFFAOYSA-Q |
Synonym | 2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt |
PubChem CID | 2723814 |
SMILES | CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10160022
|
Thermo Scientific Acros
148530250 |
25 g |
34.87 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10611643
|
Thermo Scientific Acros
148531000 |
100 g |
76.18 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10473585
|
Thermo Scientific Acros
148535000 |
500 g |
274.37 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
58052-80-5 | |
132.23 | |
MZSSRMMSFLVKPK-UHFFFAOYSA-Q | |
2723814 |
C6H18N3 | |
MFCD00149559 | |
2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt | |
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1 |
Specifications
58052-80-5 | |
100.0 | |
Beige | |
55°C | |
98% | |
C6H18N3 | |
26, I, 3 | |
Solubility in water: very soluble. Other solubilities: soluble in ethanol,chloroform | |
CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1 | |
132.23 | |
183.26 | |
Crystalline Powder |
97.5 | |
95.0°C to 97.0°C | |
110.0°C | |
Authentic | |
Glass bottle | |
MFCD00149559 | |
2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer nt | |
MZSSRMMSFLVKPK-UHFFFAOYSA-Q | |
2,4,6-trimethyl-1,3,5-triazinane;trihydrate | |
2723814 | |
98% | |
Acetaldehyde ammonia trimer, 98% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-868-2
- Acetaldehyde ammonia trimer
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only