missing translation for 'onlineSavingsMsg'
Learn More
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals Product Code.: 10160022

Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals

Product Code. 10160022
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 10160022

Brand: Thermo Scientific Acros 148530250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58052-80-5
Molecular Formula C6H18N3
Molecular Weight (g/mol) 132.23
MDL Number MFCD00149559
InChI Key MZSSRMMSFLVKPK-UHFFFAOYSA-Q
Synonym 2,4,6-trimethyl-1,3,5-triazinane trihydrate, acetaldehyde ammonia trimer, hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate, 1,3,5-triazine, hexahydro-2,4,6-trimethyl-, trihydrate, acetaldehyde ammonia trimer trihydrate, 2,4,6-trimethyl-1,3,5-triazaperhydroine, hydrate, hydrate, hydrate, acetaldehydeammoniatrimer, acmc-1bm0n, acetaldehyde-ammonia trimer trihydrate, acetaldehyde ammonia trimer ntShow MoreShow Less
PubChem CID 2723814
SMILES CC1[NH2+]C(C)[NH2+]C(C)[NH2+]1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 95.0°C to 97.0°C
Color Beige
Boiling Point 110.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 26, I, 3
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in ethanol,chloroform
IUPAC Name 2,4,6-trimethyl-1,3,5-triazinane;trihydrate
Formula Weight 183.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Acetaldehyde ammonia trimer, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457150 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number148535000,148530250,148531000
Lot NumberA0451033 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number148535000,148530250,148531000
Lot NumberA0401160 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number148535000,148530250,148531000
Lot NumberA0394539 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number148535000,148530250,148531000
Lot NumberA0412905 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number148535000,148530250,148531000
compliance-icons
Product Identifier
  • Acetaldehyde ammonia trimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetaldehyde ammonia trimer, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.