missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C
Specifications
CAS | 67-64-1 |
---|---|
Color | Colorless |
Formula Weight | 58.08g/mol |
Vapor Pressure | 247mbar at 20°C |
Viscosity | 0.32 mPaS at 20°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11433733
|
Fisher Chemical
A/0600/PB08 |
500 mL | Plastic Coated Glass bottle |
N/A
|
N/A | ||||
10395640
|
Fisher Chemical
A/0600/15 |
1 L | Amber glass bottle |
N/A
|
N/A | ||||
10162180
|
Fisher Chemical
A/0600/17 |
2.5 L | Amber glass bottle |
N/A
|
N/A | ||||
10252232
|
Fisher Chemical
A/0600/PC17 |
2.5 L | HDPE plastic bottle |
N/A
|
N/A | ||||
10324930
|
Fisher Chemical
A/0600/PB17 |
2.5 L | Plastic Coated Glass bottle |
N/A
|
N/A | ||||
10642241
|
Fisher Chemical
A/0600/21 |
5 L | HDPE plastic bottle |
N/A
|
N/A | ||||
10098181
|
Fisher Chemical
A/0600/PC21 |
5 L | HDPE plastic bottle |
N/A
|
N/A | ||||
10570521
|
Fisher Chemical
A/0600/25 |
25 L | Metal can |
N/A
|
N/A | ||||
Description
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C
Specifications
67-64-1 | |
58.08g/mol | |
0.32 mPaS at 20°C | |
7 | |
56°C |
Colorless | |
247mbar at 20°C | |
2 | |
-95°C | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.