Learn More
Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C
Brand: Fisher Chemical A/0600/15
25.94 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C

Specifications
67-64-1 | |
1 L | |
0.32 mPaS at 20°C | |
C3H6O | |
Liquid | |
7 | |
-95°C | |
CC(C)=O | |
58.08 | |
CHEBI:15347 |
Amber Glass Bottle | |
247mbar at 20°C | |
2 | |
MFCD00008765 | |
Colorless | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
56°C | |
propan-2-one | |
180 | |
58.08g/mol |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2527023 | Certificate of Analysis | 14/02/2025 | A/0600/17,A/0600/27,A/0600/PB17,A/0600/PC17,A/0600/PB08,A/0600/PC17X,A/0600/25X,A/0600/21,A/0600/15Z,A/0600/17Z,A/0600/15,A/0600/PC21,A/0600/JC21,A/0600/25SS,A/0600/25,A/0600/99SS |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.