Learn More
Cytiva AcroPak™ 1000 Capsules with Supor™ Membrane
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
Brand:  Cytiva 12994
 View more versions of this product
Product Code. 17134251
  • 231.00 € / Each
Explore more special offers
Description
Description
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
  • Designed for research applications only
  • Gamma-irradiated
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hydrophilic PES
Sterile
Stepped Hose Barb
0.8/0.45 μm
1000 cm2
1/Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva AcroPak™ 1000 Capsules with Supor™ Membrane > 0.8 μm/0.45 μm