Learn More
OHAUS™ Adventurer™ Precision Balance
Intuitive Adventurer™ Precision Balance designed for routine weighing.
Brand: OHAUS™ 30122617
1484.80 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Intuitive Balance Designed for Routine Weighing:
Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, three level user management to fulfill GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.
Unique Features Include:
- Adventurer balances feature a color touchscreen, icon-based user interface, and an ergonomic design -making them easy to configure and use.
- Features such as specialized weighing modes, multiple connectivity options, and AutoCal™ provide versatility and flexibility for a variety of applications.
- Durable construction, large weighing surfaces, a space-saving draftshield design, and full housing in-use cover allow for use in lab, education and industrial environments.
Stability, Accuracy, and Fast Operation Ensure Optimal Weighing Results in Routine Weighing Tasks:
Weighing Performance:
- Delivers stable and reliable weighing results for routine weighing tasks.
Stabilization Time:
- The fast stabilization time of the Adventurer improves productivity in the laboratory.
Calibration:
- AutoCal™ — Selected models feature the OHAUS automatic internal calibration system that performs routine maintenance by calibrating the balance daily.
- External Calibration — Traditional calibration in which the operator manually calibrates the balance with their choice of calibration weight value to ensure accuracy available on every model.
Color Touchscreen Offers Easy and Fast Operation of Adventurer Applications
- Operate and access nine Adventurer application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen.
- Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves.
- In addition to the touchscreen, Adventurer also has six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print.
Equipped with the Connectivity and Functional Features Required in Laboratories
Dual USB Ports:
- A front USB host port is easily accessible and makes it simple to load data from the balance on to a flash drive without having to reach around to the back or move the balance.
- A second USB device is located at the rear of the balance that can be used to connect the balance to a PC.
- The connectivity options help meet traceability requirements in traditional installations.
Label Printing Function:
- Easy to link with Zebra printer and have one built-in label printing template.
Balance Profiles:
- The cloning feature allows you to save user and application settings to a USB flash drive which can be easily used to configure additional Adventurer balances.
Below Minimum Sample Weight Indication:
- When using the minimum weight feature, the display clearly indicates that your current sample weight is below your defined minimum limit. Simply increase your sample weight to assure that your results are up to your standard.
Space-saving Draftshield Designed to Improve User Experience and Accessibility:
- Draftshield doors are constructed of two glass panels, reducing the space required on the lab bench when the doors are open.
- Wide door entry provides unobstructed access and allows larger weighing vessels to be easily placed on the pan, reducing the chance of accidental spillage.
- Easy to keep clean in order to ensure a safe workspace by minimizing contamination.
Power Saving Functions:
- The Adventurer is designed with various power-saving features that help reduce the environmental impact.
- These include auto-off and brightness controls.
User Management Function:
- Ensures data security and data traceability.
- 3 level user management function ensures data security and data traceability requirements.
- One administrator, two supervisors and 10 users have preset accessibility in the software.
Real Time Clock with GLP/GMP Data:
- A real-time clock function keeps accurate time even during power loss.
- GLP data capability has the ability to record Sample name, Project names and Balance IDs to help meet traceability and compliance requirements.

Specifications
Metrology: OIML R76, EN 45501 (Class I, nmax 320000, Class II, nmax 52000), Product Safety: EN 61010-1, IEC 61010-1, Electromagnetic Compatibility: IEC 61326-1, EN 61326-1 (Emissions Class B, Immunity Industrial Requirements), GLP/GMP Compliant | |
Precision Balance AX4202/E | |
Color Touchscreen | |
Weighing, Parts Counting, Percent Weighing, Check | |
±0.02 g | |
1.5 sec. | |
AX4202/E | |
3 ppm/°C | |
50°F to 86°F | |
4000 g, OIML F1 or ASTM CL 2 | |
2 years | |
354 x 230 x 100 mm | |
354 mm | |
100 mm | |
354 mm | |
12.1 lb. | |
AC Adapter Input: 100/240 V, 0.3 A, 50/60 Hz | |
AC Adapter Input: 0.3A, AC Adapter Output: 0.84A | |
No |
RS232, USB Host, USB Device | |
4200 g | |
0.01 g | |
AC Adapter | |
Metal Base, ABS Top Housing, Stainless Steel Pan | |
8.2 g | |
0.01 g | |
1.5 sec. | |
10°C to 30°C | |
Precision Balance | |
Weighing (17 units + custom unit), parts counting, percentage weighing, check weighing, animal/dynamic weighing, totalization/statistics, formulation, density determination, display hold, batch printing | |
13.93 in. | |
230 mm | |
7.7 in. | |
195 mm | |
5.5 kg | |
100/240 V | |
AC Adapter Input: 50/60 Hz | |
External |
Videos
-
Ohaus Adventurer balances
Your input is important to us. Please complete this form to provide feedback related to the content on this product.