missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Adventurer™ Precision Balance

Intuitive Adventurer™ Precision Balance designed for routine weighing.

1200.00 € - 2935.00 €

Specifications

Display Type Color Touchscreen
View More Specs

Products 36
Product Code Brand Capacity Readability Model Calibration Price Quantity & Availability  
Product Code Brand Capacity Readability Model Calibration Price Quantity & Availability  
15375043
View Documents
OHAUS™
30127960
220 g 1 mg AX223 Internal
2080.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15395043
View Documents
OHAUS™
30100623
220 g 1 mg AX223M Internal
2375.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15385043
View Documents
OHAUS™
30127961
220 g 1 mg AX223/E External
1655.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15591926
View Documents
OHAUS™
30206080
420 g 0.01 g AX422/E External
1265.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15305053
View Documents
OHAUS™
30127962
420 g 1 mg AX423 Internal
2345.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15325053
View Documents
OHAUS™
30100624
420 g 1 mg AX423M Internal
2635.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15315053
View Documents
OHAUS™
30127963
420 g 1 mg AX423/E External
1950.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15335053
View Documents
OHAUS™
30122614
520 g 1 mg AX523 Internal
2615.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15355053
View Documents
OHAUS™
30100625
520 g 1 mg AX523M Internal
2935.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15345053
View Documents
OHAUS™
30122615
520 g 1 mg AX523/E External
2245.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17941292
View Documents
OHAUS™
30805890
620 g 0.001 g AX623 Internal
2645.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
17921302
View Documents
OHAUS™
30805896
620 g 0.001 g AX623M Internal
2935.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
17991292
View Documents
OHAUS™
30805891
620 g 0.001 g AX623/E External
2245.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
15511936
View Documents
OHAUS™
30206082
820 g 0.01 g AX822/E External
1445.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15385053
View Documents
OHAUS™
30127966
1500 g 0.01 g AX1502 Internal
2035.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15305063
View Documents
OHAUS™
30100626
1500 g 0.01 g AX1502M Internal
2335.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15395053
View Documents
OHAUS™
30127967
1500 g 0.01 g AX1502/E External
1655.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15521936
View Documents
OHAUS™
30206083
2200 g 0.1 g AX2201 Internal
1715.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15531936
View Documents
OHAUS™
30206084
2200 g 0.1 g AX2201/E External
1200.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15315063
View Documents
OHAUS™
30127968
2200 g 0.01 g AX2202 Internal
2070.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15335063
View Documents
OHAUS™
30100627
2200 g 0.01 g AX2202M Internal
2355.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15325063
View Documents
OHAUS™
30127969
2200 g 0.01 g AX2202/E External
1680.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15563883
View Documents
OHAUS™
30127971
4200 g 0.1 g AX4201/E External
1445.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15345063
View Documents
OHAUS™
30122616
4200 g 0.01 g AX4202 Internal
2320.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15365063
View Documents
OHAUS™
30100628
4200 g 0.01 g AX4202M Internal
2635.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15355063
View Documents
OHAUS™
30122617
4200 g 0.01 g AX4202/E External
1950.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15375063
View Documents
OHAUS™
30122618
5.2 kg 0.01 g AX5202
2645.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15385063
View Documents
OHAUS™
30100629
5.2 kg 0.01 g AX5202M
2935.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17931292
View Documents
OHAUS™
30805892
6200 g 0.01 g AX6202 Internal
2645.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
17991282
View Documents
OHAUS™
30805897
6200 g 0.01 g AX6202M Internal
2935.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
17911302
View Documents
OHAUS™
30805893
6200 g 0.01 g AX6202/E External
2245.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
15571926
View Documents
OHAUS™
30122651
8200 g 0.1 g AX8201M Internal
2290.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15543883
View Documents
OHAUS™
30122620
8200 g 0.1 g AX8201/E External
1610.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17971282
View Documents
OHAUS™
30805894
12 kg 0.1 g AX12001 Internal
2095.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
17961302
View Documents
OHAUS™
30805898
12 kg 0.1 g AX12001M Internal
2395.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
17981282
View Documents
OHAUS™
30805895
12 kg 0.1 g AX12001/E External
1695.00 €
1 unit
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Intuitive Balance Designed for Routine Weighing:

Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, three level user management to fulfill GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.

Unique Features Include:

  • Adventurer balances feature a color touchscreen, icon-based user interface, and an ergonomic design -making them easy to configure and use.
  • Features such as specialized weighing modes, multiple connectivity options, and AutoCal™ provide versatility and flexibility for a variety of applications.
  • Durable construction, large weighing surfaces, a space-saving draftshield design, and full housing in-use cover allow for use in lab, education and industrial environments.

Stability, Accuracy, and Fast Operation Ensure Optimal Weighing Results in Routine Weighing Tasks:

Weighing Performance:

  • Delivers stable and reliable weighing results for routine weighing tasks.

Stabilization Time:

  • The fast stabilization time of the Adventurer improves productivity in the laboratory.

Calibration:

  • AutoCal™ — Selected models feature the OHAUS automatic internal calibration system that performs routine maintenance by calibrating the balance daily.
  • External Calibration — Traditional calibration in which the operator manually calibrates the balance with their choice of calibration weight value to ensure accuracy available on every model.

Color Touchscreen Offers Easy and Fast Operation of Adventurer Applications

  • Operate and access nine Adventurer application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen.
  • Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves.
  • In addition to the touchscreen, Adventurer also has six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print.

Equipped with the Connectivity and Functional Features Required in Laboratories

Dual USB Ports:

  • A front USB host port is easily accessible and makes it simple to load data from the balance on to a flash drive without having to reach around to the back or move the balance.
  • A second USB device is located at the rear of the balance that can be used to connect the balance to a PC.
  • The connectivity options help meet traceability requirements in traditional installations.

Label Printing Function:

  • Easy to link with Zebra printer and have one built-in label printing template.

Balance Profiles:

  • The cloning feature allows you to save user and application settings to a USB flash drive which can be easily used to configure additional Adventurer balances.

Below Minimum Sample Weight Indication:

  • When using the minimum weight feature, the display clearly indicates that your current sample weight is below your defined minimum limit. Simply increase your sample weight to assure that your results are up to your standard.

Space-saving Draftshield Designed to Improve User Experience and Accessibility:

  • Draftshield doors are constructed of two glass panels, reducing the space required on the lab bench when the doors are open.
  • Wide door entry provides unobstructed access and allows larger weighing vessels to be easily placed on the pan, reducing the chance of accidental spillage.
  • Easy to keep clean in order to ensure a safe workspace by minimizing contamination.

Power Saving Functions:

  • The Adventurer is designed with various power-saving features that help reduce the environmental impact.
  • These include auto-off and brightness controls.

User Management Function:

  • Ensures data security and data traceability.
  • 3 level user management function ensures data security and data traceability requirements.
  • One administrator, two supervisors and 10 users have preset accessibility in the software.

Real Time Clock with GLP/GMP Data:

  • A real-time clock function keeps accurate time even during power loss.
  • GLP data capability has the ability to record Sample name, Project names and Balance IDs to help meet traceability and compliance requirements.
Specifications

Specifications

Color Touchscreen
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.