Learn More
Welch™ Aluminum KF Inner Centering with Viton O-ring
Designed for use with CRVpro Rotary Vane Vacuum Pumps. Welch Ilmvac™ Aluminum KF Inner Centering Rings feature a replaceable Fluoroelastomer O-ring seal.
Brand:  Welch™ 701091
 View more versions of this product
Product Code. 10680343
  • 7.10 € / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel centering ring with Viton O-ring
  • Replaceable Fluoroelastomer O-ring seal
  • Centering ring assemblies are placed between two ISO flanges with matching outer diameters
  • he widest portion of the centering ring rests inside a centering ring locater groove (blank flange) or counterbore (bored flange) on the Kwik-Flange™
  • The O-ring rests on the flat polished surface outside the locater groove or counterbore
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Separator 16 DN
Aluminum
1 Each
KF Inner Centering
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Welch™ Aluminum KF Inner Centering with Viton O-ring > O.D.: 16mm