Learn More
Aluminum sulfate hydrate, 97+%, Thermo Scientific Chemicals
41.20 € - 208.00 €
Chemical Identifiers
CAS | 17927-65-0 |
---|---|
Molecular Formula | Al2O12S3 |
Molecular Weight (g/mol) | 342.13 |
MDL Number | MFCD00149136 |
InChI Key | DIZPMCHEQGEION-UHFFFAOYSA-H |
Synonym | aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o |
PubChem CID | 3080712 |
IUPAC Name | dialuminium(3+) trisulfate |
SMILES | [Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11321749
|
Thermo Scientific Alfa Aesar
012362.36 |
500 g |
41.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311749
|
Thermo Scientific Alfa Aesar
012362.A1 |
1 kg |
63.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331749
|
Thermo Scientific Alfa Aesar
012362.A7 |
5 kg |
208.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aluminum sulfate hydrate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17927-65-0 | |
342.13 | |
DIZPMCHEQGEION-UHFFFAOYSA-H | |
3080712 | |
[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Al2O12S3 | |
MFCD00149136 | |
aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o | |
dialuminium(3+) trisulfate |
Specifications
17927-65-0 | |
Crystalline | |
Al2O12S3 | |
aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o | |
DIZPMCHEQGEION-UHFFFAOYSA-H | |
dialuminium(3+) trisulfate | |
3080712 | |
Odorless | |
1.61 g/cm3 |
White | |
>97% | |
MFCD00149136 | |
It is slightly soluble in alcohol and dilute mineral acids. | |
[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
342.13 | |
342.15 (Anhydrous) | |
White crystalline | |
Aluminum sulfate hydrate, hydrate |
Safety and Handling
P280-P305+P351+P338-P310
H318
missing translation for 'einecsNumber' : 233-135-0
missing translation for 'rtecsNumber' : BD1700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum sulfate hydrate
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only