missing translation for 'onlineSavingsMsg'
Learn More
Amberlite™ IRC120 Na ion exchange resin, Thermo Scientific Chemicals Product Code.: 11368206

Amberlite™ IRC120 Na ion exchange resin, Thermo Scientific Chemicals

Product Code. 11368206
2500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11368206

Brand: Thermo Scientific Alfa Aesar L15199.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

Solubility
Insoluble in water.

Notes
Water softening, dealkalinization, deionization
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78922-04-0
Molecular Formula C13H10ClNO4S
Molecular Weight (g/mol) 311.736
MDL Number MFCD00132707
InChI Key APBOVLPLJFJSRI-UHFFFAOYSA-N
Synonym 3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid
PubChem CID 8190984
IUPAC Name 3-[(3-chlorophenyl)sulfonylamino]benzoic acid
SMILES C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O

Specifications

Merck Index 14,382
Quantity 2500 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite™ IR-120(Na) ion exchange resin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0413083 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog NumberL15199.36,L15199.0E,L15199.22
Lot NumberA0413084 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog NumberL15199.36,L15199.0E,L15199.22
Lot NumberA0452321 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberL15199.36,L15199.0E,L15199.22
Lot NumberA0413086 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberL15199.36,L15199.0E,L15199.22
Lot Number10227585 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL15199.36,L15199.0E,L15199.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amberlite™ IRC120 Na ion exchange resin, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.