missing translation for 'onlineSavingsMsg'
Learn More
Amberlite XAD-1180, Thermo Scientific Chemicals Product Code.: 11372887

Amberlite XAD-1180, Thermo Scientific Chemicals

Product Code. 11372887
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11372887

Brand: Thermo Scientific Alfa Aesar L16097.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97396-56-0
MDL Number MFCD00145830

Specifications

Merck Index 14,382
Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite XAD-1180

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10239267 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL16097.22,L16097.36
Lot Number10238810 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL16097.22,L16097.36
Lot Number10238009 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL16097.22,L16097.36
Lot Number10180639 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16097.22,L16097.36
Lot Number10129199 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16097.22,L16097.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amberlite XAD-1180, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.