missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cells-to-CT™ 1-Step Power SYBR™ Green Kit
GREENER_CHOICE

Like other members of the Cells-to-CT™ product family, the Cells-to-CT™ 1-Step Power SYBR™ Green Kit makes it possible to perform expression analysis directly from cultured cells without RNA purification.

Brand:  Invitrogen™ A25601

 View more versions of this product

Product Code. 15445840

  • 345.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2.2 mL Lysis Solution, store at 2°C to 8°C, 22 μL Dnase I, store at -5° to -30°C, 200 μL Stop Solution, store at -5° to -30°C, 500 μL Power SYBR™ Green 1-step qPCR Mix, store at -5° to -30°C, 10 μL Power SYBR™ Green 1-step RT Mix, store at -5° to -30°C
Tube
1-step RT-qPCR
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, QuantStudio™, ViiA™ 7 System, StepOnePlus™ System, StepOne™ System, Real-Time PCR Systems or Thermal Cyclers, 7500 Fast Dx System, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Cells-to-CT™, Power SYBR™ Green
20 Preps
Cell Lysis
10-100,000 Cells
SYBR
High
7 min.
Fewer resources used and less waste, less hazardous
1-Step qPCR kit
Gene Expression
Mammalian Cells
2.2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2817943 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA25601
Lot Number2807308 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA25601
Lot Number2813606 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA25601
Lot Number2825287 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA25601
Lot Number2806171 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA25601
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Cells-to-CT™ 1-Step Power SYBR™ Green Kit > 20 preps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.