missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDTA (0.5 M), pH 8.0, RNase-free

Invitrogen Molecular biology grade, 0.5 M EDTA, pH 8.0 solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use.

Brand:  Invitrogen™ AM9261

 View more versions of this product

Product Code. 10458654

  • 183.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen Molecular biology grade, 0.5 M EDTA, pH 8.0 solution is supplied in one bottle. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

EDTA
0.5 M
8
Molecular Biology Grade
Room Temperature
Store at room temperature.
Bottle
Ambion™
500 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2788590 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM9261
Lot Number2805304 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM9261
Lot Number2796670 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM9261
Lot Number2833365 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM9261
Lot Number2848908 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM9261
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ EDTA (0.5 M), pH 8.0, RNase-free > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.