missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GlycoBlue™ Coprecipitant (15 mg/mL)

GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant.

Brand:  Invitrogen™ AM9515

 View more versions of this product

Product Code. 10391565

  • 183.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant. The attached dye increases visibility of the pellet. This product is an ideal coprecipitant in nuclease protection assays at 1/100 dilution of stock solution.

Features of GlycoBlue™ Coprecipitant:
• Ideal for RT-PCR
• Increases pellet mass and visibility
• Quantitative recovery of low concentrations (ng/mL) of nucleic acid
• Prevents pellet loss in nuclease protection assays

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

15 mg/mL
Tube
Molecular Biology Grade
Dry Ice
Store at –20°C.
Ambion™, GlycoBlue™
300 μL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2786427 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM9515
Lot Number2825548 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM9515
Lot Number2865271 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM9515
Lot Number2865280 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberAM9515
Lot Number3057382 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberAM9515
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ GlycoBlue™ Coprecipitant (15 mg/mL) > 300μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.