missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Retic Lysate IVT™ Kit

Efficiently translates high molecular weight proteins and both capped and uncapped mRNAs

Brand:  Invitrogen™ AM1200

 View more versions of this product

Product Code. 10137194

  • 477.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Retic Lysate should be stored at -80°C. 20X Translation Mix minus-Met, 20X Translation Mix minus-Leu, High Salt Mix minus-Met, Low Salt Mix minus-Met, and Capped Xef-1 RNA should be stored below -70°C. Nuclease-free Water can be stored at any temperature.
This item is not returnable. View return policy

Description

Description

The Retic Lysate IVT™ Kit efficiently translates in vitro-synthesized transcripts, poly(A) RNA, and total RNA, including difficult-to-translate RNAs. The optimized lysate results in high protein yield, biological activity, and consistent performance. A report in Nucleic Acids Research (1) found that some commercial reticulocyte lysates had poor fidelity for initiation at the appropriate AUG initiator codon. The Retic Lysate IVT Kit was tested using Kozak's constructs and it demonstrated high fidelity for initiating at the proper AUG.

  • Efficiently translates a wide variety of high molecular weight proteins and both capped and uncapped mRNAs
  • Accommodates large volume of template mRNA
How it works
The lysate has been optimized for translation by the addition of an ATP-regenerating system, hemin, and calf liver tRNA. Included in the kit is an optimized Translation Mix (-Met or -Leu) for the translation of both capped and uncapped messages along with High and Low Salt buffers. These buffers can be used to maximize protein yield and activity from difficult-to-translate sequences.

1. Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res 18:2828.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 5 × 200 μl (store at -80°C)
• 2 × 1.75 ml Nuclease-free Water (store at any temperature)
• 100 μl 20X Translation Mix minus-Met
• 100 μl 20X Translation Mix minus-Leu
• 100 μl High Salt Mix minus-Met
• 100 μl Low Salt Mix minus-Met
• 50 μl Capped Xef-1 RNA (50 μg/ml)

Unless otherwise stated, store reagents below -70°C. Do not store cell-free translation systems in the liquid phase of a liquid nitrogen storage system.
Water Soluble Protein
Cell-Free
60 Reactions
Retic Lysate
Dry Ice
Kit
Cell-Free Expression
In Vitro Transcription
Ambion™
60 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2821365 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1200
Lot Number2821373 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1200
Lot Number2829732 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1200
Lot Number2821375 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1200
Lot Number2807682 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM1200
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Retic Lysate IVT™ Kit > 60 reactions; Without Manual

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.