Learn More
Thermo Scientific™ Tango™ EDG4-bla U2OS Cells
The Tango™ EDG4-bla U2OS cells contain the human Endothelial Differentiation Gene 4 (EDG4) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand:  Thermo Scientific™ K1442
Product Code. 10721824
  • 13350.00 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Tango™ EDG4-bla U2OS cells contain the human Endothelial Differentiation Gene 4 (EDG4) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ EDG4-bla U2OS DA cells have been functionally validated for Z' factor and EC50 concentrations of LPA (18:1). In addition, Tango™ EDG4-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1458354 Certificate TypeCertificate of Analysis Date16/04/2017 Catalog NumberK1442
Lot Number40501 Certificate TypeCertificate of Analysis Date16/04/2017 Catalog NumberK1442
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Tango™ EDG4-bla U2OS Cells > 2 x 106 Cells