Learn More
Cytiva Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System
Use for fast and easy detection of medium-to-high-target amounts. Cytiva Life Sciences™ Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction
Brand:  Cytiva RPN2105
 View more versions of this product
Product Code. 10195804
  • 772.00 € / Each
Explore more special offers
Includes: 45-000-922 and 45-000-923: Labeling reagent; Crosslinker; Control DNA; Blocking agent; ECL detection reagents and ECL Gold hybridization buffer
Description
Description
    • Direct probe labeling in a 10 min reaction, 1 h from hybridization to detection with ECL Direct, Hybond N+, and Hyperfilm ECL.
    • Eliminates handling, waste, and regulatory issues associated with the use of radioactivity.
    • No need to strip blots before reprobing.
    • For fast and easy detection of medium- to high-target amounts in applications such as colony/plaque screens, dot blots, and PCR product analyses.
    • Consistent results combining strong signals with very low backgrounds.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2°C to 8°C
ECL Detection Reagents 1 and 2, for 4000cm2 membrane
For 4000 cm2 membrane
Colony or plaque screens, dot blots and PCR product analyses
ECL Detection Reagents
20 min.