missing translation for 'onlineSavingsMsg'
Learn More
Ammonium hydroxide, for HPLC, 35% solution in water, Thermo Scientific Chemicals Product Code.: 15547049

Ammonium hydroxide, for HPLC, 35% solution in water, Thermo Scientific Chemicals

Product Code. 15547049
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 15547049

Brand: Thermo Scientific Acros 460801000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1336-21-6, 7732-18-5
Molecular Formula H5NO
Linear Formula NH4OH
MDL Number MFCD00066650
Density 0.8800g/mL
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
Solubility Information Solubility in water: soluble.
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 100 mL
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Formula Weight 35.05
Grade HPLC
Specific Gravity 0.88
Chemical Name or Material Ammonium hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2421088 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number460801000
Lot Number2401832 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number460801000
Lot Number2409127 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number460801000
Lot Number1918967 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number460801000
Lot Number1989802 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number460801000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide 35%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium hydroxide, for HPLC, 35% solution in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.