Learn More
Aniline Point Standard, ASTM D 611 (Nominal Value: 62.30 °C ), Paragon Scientific™
Fuels Testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Aniline Point ASTM D611
Brand: Paragon Scientific CRM-APKR
Description
- Certified Reference Material for Aniline Point
- Manufactured in accordance with ASTM / EN or IP test methods
- Certified via Round Robin
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
250 mL | |
UN 1223 |
Nominal values quoted. Certified Values will vary from batch to batch.