Learn More
Fisherbrand™ AP Bio 1000 Air Purification Unit
The unique AP BIO 1000 Air Purifier is the best and proven way to combat airborne transmission of viruses and bacteria in almost any closed environment.
Brand:  Fisherbrand™ AP10BSCW00
Product Code. 17428982
  • 3965.00 € / Each
Explore more special offers
Description
Show More
Specifications
Specifications
200 to 1000 m3/h
66 Kg
2 ft. 5 9⁄64 in.
435 mm
Air Purifier
145.5 lb.
1 ft. 5 1/8 in.
1245 mm
740 mm
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ AP Bio 1000 Air Purification Unit > Sample Flow Rate: 200 to 1000 m3/h; Weight (English): 145.5 lb.; Weight (Metric): 66 kg; Width (Metric): 740 mm