missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG

TaqMan™ Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.

Brand:  Applied Biosystems™ 4366073

 View more versions of this product

Product Code. 10575385

  • 1784.00 € / 1250 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Template and primers
This item is not returnable. View return policy

Description

Description

  • Results in 40 minutes for 40 cycles of PCR in a 20 μL reaction volume when using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems
  • The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup
  • A proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments
  • Contains everything needed for TaqMan-based qPCR amplification and detection in a single-tube format
  • TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR
  • Compatible with TaqMan Gene Expression assays
  • Store at 2°C to 8°C

Order Info

Guaranteed minimum shelf life is 60 days.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

This 5-pack contains five kits, each containing 2 tubes (1.25 ml each) of 2X TaqMan™ Fast Universal PCR Master Mix.

Store at 2°C to 8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Tube
qPCR
Fast
ROX (Pre-mixed)
Fast Universal PCR Master Mix
250 Reactions
2X
1250 Reactions
1.25 mL
Primer-probe
High
AmpliTaq DNA Polymerase
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
AmpErase™, TaqMan™
1,250 reactions
Gene Expression, Genotyping, miRNA analysis
2 X
DNA (Genomic), cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.