Learn More
Applied Biosystems™ TaqMan™ Fast Universal PCR Master Mix (2X), no AmpErase™ UNG
TaqMan™ Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR.
380.00 € - 6980.00 €
Specifications
Detection Method | Primer-probe |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq DNA Polymerase |
Requires: Template and primers
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10311135
|
Applied Biosystems™
4366072 |
500 Reactions |
774.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10637755
|
Applied Biosystems™
4367846 |
5000 Reactions |
6980.00 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10116674
|
Applied Biosystems™
4352042 |
250 Reactions |
380.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10575385
|
Applied Biosystems™
4366073 |
1250 Reactions |
1820.00 €
1250 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10702697
|
Applied Biosystems™
4364103 |
2500 Reactions |
3545.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Results in 40 minutes for 40 cycles of PCR in a 20 μL reaction volume when using the Applied Biosystems 7900HT and Fast 7500 Fast real-time PCR systems
- The optimized formulation contains AmpliTaq Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start, minimizing non-specific product formation and allowing room temperature reaction setup
- A proprietary ROX dye serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations, for superb precision on Applied Biosystems real-time PCR instruments
- Contains everything needed for TaqMan-based qPCR amplification and detection in a single-tube format
- TaqMan Fast Universal PCR Master Mix contains all of the components, excluding the template and primers, for superior real-time qPCR
- Compatible with TaqMan Gene Expression assays
- Store at 2°C to 8°C
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
Primer-probe | |
High | |
AmpliTaq DNA Polymerase | |
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode | |
AmpErase™, TaqMan™ | |
Gene Expression, Genotyping, miRNA analysis | |
2 X |
Tube | |
qPCR | |
Fast | |
ROX (Pre-mixed) | |
Fast Universal PCR Master Mix | |
2X | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.