missing translation for 'onlineSavingsMsg'
Learn More

Extract All DNA Extract All Reagents Kit

Provides PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues, in 5 minutes

188.00 € - 4745.00 €

Specifications

Content And Storage Store at 2–8°C.
Product Line Extract All
Reagent Type Cell Lysis Buffer
Test Time 5 min
Form Liquid
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11571296
View Documents Promotion Details
Extract All
4403319
5 mL
188.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11561296
View Documents Promotion Details
Extract All
4402616
20 mL
718.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11551296
View Documents Promotion Details
Extract All
4402599
200 mL
4745.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Simply mix your sample with the DNA lysis solution provided. Incubate for 3 minutes and add the DNA stabilizing solution, also provided.

After extraction, robust PCR amplification can be completed using TaqMan™ GTXpress™ Master Mix or TaqMan™ OpenArray™ Genotyping Master Mix (pre-amplified sample only), depending on your application. PCR can be performed using either Fast or Standard mode thermal cycling conditions. The DNA Extract All Reagents Kit is included in the TaqMan Sample-to-SNP Kit, so workflow details can be found in the TaqMan™ Sample-to-SNP user guide.

Thermal cycling conditions should be followed according to the master mix being used. TaqMan™ GTXpress™ Master Mix is required for PCR amplification of sample prepared using the DNA Extract All Reagents Kit. If sample has undergone preamplification, OpenArray™ Genotyping Master Mix may also be used.

Recommended Storage

Store at 2° to 8°C.

Specifications

Specifications

Store at 2–8°C.
Cell Lysis Buffer
Liquid
Extract All
5 min
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DNA Extract All Reagents Kit

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.