Learn More
Extract All DNA Extract All Reagents Kit
Provides PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues, in 5 minutes
Brand: Extract All 4403319
124.74 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Simply mix your sample with the DNA lysis solution provided. Incubate for 3 minutes and add the DNA stabilizing solution, also provided.
After extraction, robust PCR amplification can be completed using TaqMan™ GTXpress™ Master Mix or TaqMan™ OpenArray™ Genotyping Master Mix (pre-amplified sample only), depending on your application. PCR can be performed using either Fast or Standard mode thermal cycling conditions. The DNA Extract All Reagents Kit is included in the TaqMan Sample-to-SNP Kit, so workflow details can be found in the TaqMan™ Sample-to-SNP user guide.
Thermal cycling conditions should be followed according to the master mix being used. TaqMan™ GTXpress™ Master Mix is required for PCR amplification of sample prepared using the DNA Extract All Reagents Kit. If sample has undergone preamplification, OpenArray™ Genotyping Master Mix may also be used.
Recommended Storage
Store at 2° to 8°C.

Specifications
Store at 2–8°C. | |
5 mL | |
5 min |
Extract All | |
Cell Lysis Buffer | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- DNA Extract All Reagents Kit
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2810178 | Certificate of Analysis | 29/01/2025 | 4403319 |
2789518 | Certificate of Analysis | 29/01/2025 | 4403319 |
2832614 | Certificate of Analysis | 29/01/2025 | 4403319 |
2838989 | Certificate of Analysis | 29/01/2025 | 4403319 |
2838994 | Certificate of Analysis | 29/01/2025 | 4403319 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.