Learn More
Extract All DNA Extract All Reagents Kit
Provides PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues, in 5 minutes
Brand: Extract All 4402616
476.28 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Simply mix your sample with the DNA lysis solution provided. Incubate for 3 minutes and add the DNA stabilizing solution, also provided.
After extraction, robust PCR amplification can be completed using TaqMan™ GTXpress™ Master Mix or TaqMan™ OpenArray™ Genotyping Master Mix (pre-amplified sample only), depending on your application. PCR can be performed using either Fast or Standard mode thermal cycling conditions. The DNA Extract All Reagents Kit is included in the TaqMan Sample-to-SNP Kit, so workflow details can be found in the TaqMan™ Sample-to-SNP user guide.
Thermal cycling conditions should be followed according to the master mix being used. TaqMan™ GTXpress™ Master Mix is required for PCR amplification of sample prepared using the DNA Extract All Reagents Kit. If sample has undergone preamplification, OpenArray™ Genotyping Master Mix may also be used.
Recommended Storage
Store at 2° to 8°C.

Specifications
Store at 2–8°C. | |
20 mL | |
5 min |
Extract All | |
Cell Lysis Buffer | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2794272 | Certificate of Analysis | 29/01/2025 | 4402616 |
2817477 | Certificate of Analysis | 29/01/2025 | 4402616 |
2839839 | Certificate of Analysis | 29/01/2025 | 4402616 |
2839842 | Certificate of Analysis | 29/01/2025 | 4402616 |
2852990 | Certificate of Analysis | 28/01/2025 | 4402616 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.