missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Fast Virus 1-Step Master Mix for qPCR

TaqMan Fast Virus 1-Step Master Mix is designed for fast, highly sensitive real-time RT-PCR, even in the presence of challenging PCR inhibitors.

Brand:  Applied Biosystems™ 4444432

 View more versions of this product

Product Code. 13498446

  • 464.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2796636 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4444432
Lot Number2751808 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4444432
Lot Number2820154 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4444432
Lot Number2763426 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4444432
Lot Number2784388 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4444432
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ TaqMan™ Fast Virus 1-Step Master Mix for qPCR > 200 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.