missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Optical Adhesive Film

Reduces chance of well-to-well contamination and sample evaporation when applied to a microplate

Brand:  Applied Biosystems™ 4311971

245.34 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 10299204

  • 368.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price


Includes: 100 Optical Adhesive Covers
This item is not returnable. View return policy

Description

Description

Applied Biosystems MicroAmp Optical Adhesive Film is designed to reduce well-to-well contamination and sample evaporation. The easy-to-use film is applied by peeling away the backing and firmly pressing the adhesive film across the top of a microplate. This tight seal will help ensure consistent real-time PCR data without obscuring samples during reading, and the protective covering allows microplates to be more easily stacked for space-saving convenience and storage.

Features

  • Pressure-sensitive adhesive provides a tight seal over every well
  • Will not interfere with sample reading
  • Easy-to-apply film will not stick to gloves
  • Prevents sample loss and reduces cross-contamination
  • Designed to fit MicroAmp 96- and 384-well plates

Notes

Thermo Fisher Scientific has performed exhaustive functional testing regarding minor visual inconsistencies (e.g., wrinkles, line impressions, haziness, white spots, etc.) of the MicroAmp Optical Adhesive Film that may result from removal of the liner or application to a plate. These minor visual inconsistencies do not affect the performance of the adhesive tape. Should you have any additional questions, please contact Technical Support.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Optical
96
100 covers
2720 Thermal Cycler, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ViiA™ 7 Dx Fast System, QuantStudio™ Dx, 7500 Fast Dx System, ViiA™ 7 Fast System, ProFlex™ PCR System
Film, Seal, or Applicator
Pressure Sensitive
Polyester
100 Covers
Includes: 100 Optical Adhesive Covers
Storage: Room temperature
Room Temperature
MicroAmp
96/384-well Plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number33RDWW Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number4311971,4311971_LITL18 PROMO
Lot Number33P856 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number4311971,4311971_LITL18 PROMO
Lot Number33NRYR Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number4311971,4311971_LITL18 PROMO
Lot Number33PANJ Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number4311971,4311971_LITL18 PROMO
Lot Number33TN88 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number4311971,4311971_LITL18 PROMO
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ MicroAmp™ Optical Adhesive Film > 100 covers optical

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.