missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ TaqMan™ Gene Expression Master Mix

Precise and reliable real-time qPCR quantification over 9 logs of linear dynamic range with Applied Biosystems™ TaqMan™ Gene Expression Master Mix.

Brand:  TaqMan™ 4369016

297.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10525395

  • 594.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Template and primers
This item is not returnable. View return policy

Description

Description

TaqMan™ Gene Expression Master Mix has been validated with TaqMan™ Gene Expression Assays and Applied Biosystems™ real-time systems, providing precise quantification for a variety of gene expression-based real-time qPCR applications, including rare transcript detection, gene expression analysis, pathogen detection and viral load quantification,co-amplification of two targets, as well as specific detection of gene family members

TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure) for hot start activation and improved detection of bacterial targets, a blend of dNTPs with dTTP/dUTP and Uracil-DNA Glycosylase (UDG) to minimize carry-over PCR contamination, and a passive internal reference based on proprietary ROX™ dye for superb precision on Applied Biosystems™ real-time PCR instruments.

  • Provides precise and reliable real-time qPCR quantification over nine logs of linear dynamic range
  • TaqMan Gene Expression Master Mix is an optimized 2X mix that contains all of the components, excluding the template and primers, for sensitive detection down to one copy of target
  • Extended benchtop stability provides consistent results for automated liquid-handling systems
  • This makes it the choice for high-throughput sample processing overnight or over the weekend
  • Validated with TaqMan gene expression assays and Applied Biosystems real-time systems, providing precise quantification for a variety of gene expression–based real-time qPCR applications
  • Applications include rare transcript detection, gene expression analysis, pathogen detection and viral load quantification, co-amplification of two targets, as well as specific detection of gene family members
  • Store at 2°C to 8°C

TaqMan Gene Expression Master Mix contains:

  • AmpliTaq Gold DNA Polymerase, UP (Ultra Pure), for hot start activation and improved detection of bacterial targets
  • A blend of dNTPs with dTTP/dUTP and uracil-DNA glycosylase (UDG) to minimize carry-over PCR contamination
  • A passive internal reference based on proprietary ROX dye for superb precision on Applied Biosystems real-time PCR instruments

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 bottle (5 mL) 2X TaqMan™ Gene Expression Master Mix sufficient for 200 reactions. Store at 2°C to 8°C.


TaqMan™ Gene Expression Master Mix contains AmpliTaq Gold™ DNA Polymerase (Ultra Pure), Uracil-DNA glycosylase, dNTPs (with dUTP), ROX™ Passive Reference, and optimized buffer components.
Liquid
AmpliTaq Gold DNA Polymerase
7000 System, 7300 System, 7500 System, 7900HT System, Applied Biosystems StepOnePlus™ Fast Real-Time PCR SystemStepOne™, Standard Mode, StepOnePlus™, Standard Mode
TaqMan™
1 x 5 mL
200 Reactions
1 x 5 mL
Primer-probe
qPCR
Standard
ROX (Pre-mixed)
UP (Ultra Pure)
Gene Expression
DNA (Genomic), cDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.