missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Multiplex Master Mix
Applied Biosystems™ TaqMan™ Multiplex Master Mix is formulated to amplify up to 4 targets in a single reaction, delivering shorter run times (less than 40 minutes).
155.00 € - 5100.00 €
Specifications
Detection Method | Primer-probe |
---|---|
GC-Rich PCR Performance | High |
PCR Method | Multiplex qPCR |
Polymerase | AmpliTaq Fast DNA Polymerase |
Reaction Speed | Fast |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15480614
|
Applied Biosystems™
4461881 |
1 mL |
155.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15529001
|
Applied Biosystems™
4461882 |
5 mL |
692.00 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15441823
|
Applied Biosystems™
4461884 |
2 x 5 mL |
1254.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15564279
|
Applied Biosystems™
4484262 |
5 x 5 mL |
2790.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15362326
|
Applied Biosystems™
4484263 |
10 x 5 mL |
5100.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420624
|
Applied Biosystems™
4486295 |
50 mL |
5020.00 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Optimized for multiplexing - validated for quadplex gene expression and genotyping assays
- Best-in-class performance - superior sensitivity, accuracy, dynamic range, and specificity
- Reduced run times on fast and standard instrumentation - optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
- Seamless integration into your experiments - validated with Applied Biosystems™ TaqMan™ assays and TaqMan™ QSY™ probes for gene expression and genotyping experiments
- Carry-over contamination control - contains heat-labile UDG
- Broad instrument compatibility
Specifications
Primer-probe | |
Multiplex qPCR | |
Fast | |
Multiplex Master Mix | |
2X | |
DNA |
High | |
AmpliTaq Fast DNA Polymerase | |
TaqMan™ | |
Gene Expression, Virus Detection | |
2 X |
For Research Use Only. Not for use in diagnostic procedures.