missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Multiplex Master Mix

Applied Biosystems™ TaqMan™ Multiplex Master Mix is formulated to amplify up to 4 targets in a single reaction, delivering shorter run times (less than 40 minutes).

Brand:  Applied Biosystems™ 4461881

62.00 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15480614

  • 155.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Optimized for multiplexing - validated for quadplex gene expression and genotyping assays
  • Best-in-class performance - superior sensitivity, accuracy, dynamic range, and specificity
  • Reduced run times on fast and standard instrumentation - optimized for fast mode on fast instruments and fast cycling conditions on standard instruments
  • Seamless integration into your experiments - validated with Applied Biosystems™ TaqMan™ assays and TaqMan™ QSY™ probes for gene expression and genotyping experiments
  • Carry-over contamination control - contains heat-labile UDG
  • Broad instrument compatibility
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 1 tube (1 mL) of 2X TaqMan™ Multiplex Master Mix sufficient for 100 reactions.


TaqMan™ Multiplex Master Mix contains AmpliTaq™ Fast DNA Polymerase, uracil-N glycosylase (UNG), dNTPs (with dUTP), MUSTANG PURPLE™ dye (passive reference), and optimized buffer.

Store in refrigerator (2–8°C).

High
AmpliTaq Fast DNA Polymerase
7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System
Multiplex Master Mix
100 Reactions
2X
100 Reactions
1 mL
Primer-probe
Multiplex qPCR
Fast
TaqMan™
1 mL
Gene Expression, Virus Detection
2 X
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.