Learn More
3M™ Attest™ Biological Indicator
3M™ Attest™ Biological Indicator monitors the effectiveness of the steam sterilization process and provide results within 48 hours.
Brand: 3M™ 70200503483
This item is not returnable.
View return policy
Description
- Biological indicator vials are easy to use and interpret with visual color readout in 48 hours
- Attest Biological Indicator 1262 for 250°F/121°C Gravity and 270°F/132°C Vacuum Assisted Steam Sterilizers
- Use in conjunction with 3M™ Attest™ Biological Indicator Incubator 116
- The self-contained BI for 250°F/121°C gravity and 270°F/132°C vacuum-assisted steam cycles consist of a Geobacillus stearothermophilus spore strip; sealed glass ampule with growth medium and a bromocresol purple pH indicator system; brown color-coded cap with holes for sterilant penetration and a filter that acts as a barrier, and a process indicator on the label that changes from rose to brown when processed. After sterilization, the vial is crushed to join the growth media with the processed spore strip. The BI is incubated for 48 hours for a visual change readout. A color change to yellow indicates surviving spores and a positive result.
Specifications
1262 | |
Brown Cap | |
Steam |
Biological Indicator | |
100/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
3M™ Attest™ Biological Indicator > Sterilization Method: Steam
Spot an opportunity for improvement?Share a Content Correction