Learn More
Kartell Automatic pipette washer
BIOPHARMA
Ideal quick and safe solution for washing pipettes up to 60mm long. Kartell™ Automatic Pipette Washer is a high speed way of washing long pipettes that eliminates backsymphoning, as the water inlet is located above and away from any contaminated waters.
Brand:  Kartell 0021700
Product Code. 10040991
  • 229.84 € / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High quality and high speed
  • Solves the problem of washing pipettes up to 600mm long quickly and safely
  • Indispensable in the laboratory where radioactive, caustic and toxic materials are in use
  • The PE rinser fills via the spray nozzle situated on the top rim
  • This eliminates backsymphoning as the water inlet is located above and away from any contaminated waters
  • Contaminated waters will syphon out totally, once the syphon point at the top of the rinser has been reached, via the large diameter drain situated at lowest point, before refilling automatically
  • During washing the pipettes are housed in specially designed baskets which minimize breakages as it buffers the tips of pipettes against the soft PE base
Specifications
Specifications
Polyethylene, Polypropylene
Automatic Pipette Washer
734 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT24I01456 Certificate TypeDeclaration of Conformity Date09/01/2025 Catalog Number0021700
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Kartell Automatic pipette washer >