Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
GREENER_CHOICE
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Brand:  Thermo Scientific™ 904-253
543.16 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
 View more versions of this product
Product Code. 10565051
  • 627.00 € / Pack of 4800
Estimated Shipment: 23-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
  • Available nonsterile with-or-without Thermo Scientific™ ART™ filter technology
  • Available in filtered, low retention or extended length for increased volume to reach into deep well plates
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 μL
Non-sterile
Autoclavable
Free of RNase, DNase, DNA, ATP, and endotoxin
Conductive
Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Non-filtered
Black
ISO 9001 and ISO 13485
Rack
4800 Tips
Certified Quality
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms > Automation tips for Tecan™ Volume: 1000μL; Sterility: Non-sterile; Color: Conductive; Unit Size: 50 blisters of 96 tips (4800 tips in total)