missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Avidin
Avidin
Brand: Novex™ 434401
107.92 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
Reactivity: Avidin reacts with biotin.
Specifications
Unconjugated | |
Store in refrigerator (2–8°C). | |
Lyophilized |
10 mg | |
Wet Ice |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Avidin (from Egg White)
Version: Rev 1.0 (Mar 2013)
File size: 50.4 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novex™ Avidin > 10mg
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction