missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

For sample collection, in-vitro growth chambers and long term storage.

Brand:  Axygen™ P-96-450V-B

102.35 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 12587917

  • 137.03 € / Pack of 50
Estimated Shipment: 29-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    ANSI, SLAS 1-2004
    8 x 12
    Blue
    550 μL
    Non-sterile
    Sample collection, in-vitro growth chambers and long term storage
    -112°F
    Standard
    Autoclavable
    Polypropylene
    96
    Conical Bottom (V-well)
    4000 x g
    -80°C
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Axygen™ Storage Microplates > 96-well assay; V-bottom plate; Blue; 500μL

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.