missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
BIOPHARMAGREENER_CHOICE

For sample collection, in-vitro growth chambers and long term storage.

Brand:  Axygen™ P-2ML-SQ-C

106.21 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.


Product Code. 11360105

  • 132.00 € / Pack of 25
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
 

  • 2.2 mL deep well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film

Currently experiencing high demand for this item. Posted availability date may not be current. Contact Customer Support for up-to-date availability.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANSI, SLAS 1-2004
Without Lid
Clear
96
Square
25/Cs.
-112°F
Standard
8 x 12
Polypropylene
Non-sterile
Sample collection, in-vitro growth chambers and long term storage
5/Pk.
-80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number33424012 Certificate TypeCertificate of Quality Date11/04/2025 Catalog NumberP-2ML-SQ-C
Lot Number33324021 Certificate TypeCertificate of Quality Date11/04/2025 Catalog NumberP-2ML-SQ-C
Lot Number31824009 Certificate TypeCertificate of Quality Date27/03/2025 Catalog NumberP-2ML-SQ-C
Lot Number30424027 Certificate TypeCertificate of Quality Date25/03/2025 Catalog NumberP-2ML-SQ-C
Lot Number30724032 Certificate TypeCertificate of Quality Date18/03/2025 Catalog NumberP-2ML-SQ-C
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ Storage Microplates > 96-well; 2.2 mL; No; Square

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.