missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Polypropylene Measuring Scoop
Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.
Brand: Azlon SWL006
This item is not returnable.
View return policy
Description
Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.
- Made from food safe polypropylene
- Top is flat for levelling off powders
- Base is flat enabling scoops to stand horizontally
- The capacity is clearly molded on each size

Specifications
Measuring Scoop | |
Handle | |
82 mm |
Polypropylene | |
5 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product information
File size: 2 MB
Product information
File size: 3 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Azlon™ Polypropylene Measuring Scoop > Capacity: 5mL; Length: 82mm
Spot an opportunity for improvement?Share a Content Correction