missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Multi Thief

Enables the sample volume to be quickly and easily changed by changing the removable sample cells. Sampling Systems™ Multi Thief is ideal for taking multiple bulk samples from free flowing powders and granules.

Brand:  Sampling Systems™ 1290K-1000

 View more versions of this product

Product Code. 15829241

  • 815.00 € / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 316 stainless steel construction
  • Variable sample volumes
  • Range of sampler lengths
  • Suitable for free flowing powders and granules
  • Multilevel Powder Sampler with variable Sample Volumes
Note: 3 inserts are required. Inserts are not included; sold separately.

Specifications

Specifications

Multilevel Powder Sampler
Stainless Steel (316L)
40 mm
1000 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sampling Systems™ Multi Thief > LengthMetric: 1000mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.