missing translation for 'onlineSavingsMsg'
Learn More

Synergy Healthcare™ Azo™ Gusseted Reels

High quality preformed, heat sealable, peelable sterile barrier packaging. Synergy Healthcare™ Azo™ Gusseted Reels are suited to the sterilization of surgical instruments, laboratory equipment or small to medium sized sets using steam or Ethylene Oxide.

Brand:  Synergy Healthcare™ RG15050100

 View more versions of this product

Product Code. 12390479

This item is not returnable. View return policy

Description

Description

  • Protects its contents from deterioration, damage and contamination
  • Meet EN ISO 11607 part 1 and EN 868-4 standards
Specifications

Specifications

Sterilizer Bag
EN868-5 and ISO11607-1
Non-sterile
Steam and EO
100 m
Autoclavable
Blue
4/Cs.
100 m x 150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Synergy Healthcare™ Azo™ Gusseted Reels > Width: 200mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.