Learn More
Sartorius Balance Printers
Transfer data automatically or manually. Sartorius Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
Brand: Sartorius YDP10-0CE
869.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Transfer data automatically or manually. Sartorius™ Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
- Built-in real-time clock
- Built-in statistics program
- Counter for consecutive numbering of transactions can be activated
- High contrast LCD shows date or time as well as current transaction number
- Integrated rechargeable batteries with battery level indicator
- Ultrafast print head
- Runs on the balance's power; no separate power supply necessary
- Easy to change paper roll and ink ribbon cassette
Specifications
CPA Balance, Extended Balance, Moisture Analyzers. (For use w/ Cubis 1 MSE or MSA models.) | |
230 mm | |
80 mm | |
0.77 kg |
230 x 158 x 80 mm | |
158 mm | |
Interface cable, Paper roll, 4 rechargeable batteries | |
Data Printer |