missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium crystalline dendritic solid, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Silver to Gray |
Appearance | Silver Gray solid |
Assay Percent Range | 99.9% (Metals basis) |
Quantity | 5 g |
Odor | Odorless |
Form | Crystalline Dendritic Solid |
Chemical Name or Material | Barium |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X27H023 | Certificate of Analysis | 12/04/2022 | 014454.14,014454.06 |
X27H024 | Certificate of Analysis | 12/04/2022 | 014454.14,014454.06 |
B04P20 | Certificate of Analysis | 08/12/2021 | 014454.14,014454.06 |
T21G073 | Certificate of Analysis | 08/12/2021 | 014454.14,014454.06 |
T27H003 | Certificate of Analysis | 08/12/2021 | 014454.14,014454.06 |
Safety and Handling



Product Identifier
- Barium crystalline dendritic solid
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 1
- H228-Flammable solid.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- MIXTURE LIST-Contains : Barium
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Barium crystalline dendritic solid, 99.9% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction