Learn More
Buerkle™ Barrel Valve
Enables quick and clean emptying even with highly viscous liquids. Suitable for metal barrel with 2 in. BSP TriSure metal barrel screw joint.
Brand:  Buerkle™ 0536-0010
Product Code. 15655494
  • 163.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Opens, empties and recloses the barrel closure even with lying, full barrels without having to move them.
  • Fiberglass reinforced Polypropylene
  • NBR Seals
Specifications
Specifications
Blue
Barrel with 2 in. BSP TriSure
Glas fiber einforced Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Barrel Valve > Fiberglass reinforced Polypropylene