Learn More
BD Horizon™ BV421 Annexin V
A sensitive probe for identifying apoptotic cells, binding to negatively charged phospholipid surfaces with a higher affinity for phosphatidylserine (PS) than most other phospholipids
Brand:  BD 563973
526.25 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15863249
  • 530.00 € / Each
Explore more special offers
Specifications
Flow Cytometry
BV421
Buffered Solution
100 Tests
Store undiluted at 4°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Horizon™ BV421 Annexin V > 100 Tests