missing translation for 'onlineSavingsMsg'
Learn More

Lysing Buffer, BD

Buffered ammonium chloride-based lysing reagent

Brand:  BD Biosciences 555899

Product Code. 15838518

  • 209.04 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Concentration: 10x
  • Lyses red blood cells following monoclonal antibody staining on dilution
  • Maintains leukocytes viability after red blood cell lysis
  • Lysing solution results in good light scatter separation of lymphocytes, RBC debris by flow cytometry

Flow cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Liquid
RUO
4°C
100mL
Lysis Buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lysing Buffer, BD > 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For research use only.