missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Biohazard Incinerator Disposal Carton
Includes polyethylene liner. Bel Art™ Biohazard Incinerator Disposal Cartons are clearly marked for proper handling and processing.
Brand: Bel-Art™ F13205-0001
This item is not returnable.
View return policy
Description
- Corrugated cardboard carton features fitted lid with flap
- Lined with clear 1.5 mil polyethylene biohazard bag that resists tears, punctures
- Bright biohazard markings for immediate and continuous identification
- Instructions in English, Spanish, French and German
- Use with Bel-Art reusable cover for biohazard carton (Mfr. No. F132040000)
- Dimensions 12 x 12 x 27 in.
Specifications
14.4 gal. | |
12 x 12 x 27 in. (30.5 x 30.5 x 68.6 cm) | |
30.5 cm | |
Not Autoclavable | |
Proper handling and disposal of biohazard waste | |
12 in. | |
27 in. | |
Biohazard Incinerator Carton Container |
Red, White | |
68.6 cm | |
30.5 cm | |
Polyethylene | |
12 in. | |
1.5 mm | |
Cardboard |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ Biohazard Incinerator Disposal Carton > Incinerator carton; 30.4 x 30.4 x 68.6cm
Spot an opportunity for improvement?Share a Content Correction