missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Cleanware™ Self-Adhesive Waste Bags

Disposable waste bags free bench or hood space when hung from walls. Bel Art™ Cleanware™ Self-Adhesive Waste Bags adhere to cabinets and other surfaces.

Brand:  Bel-Art™ F13174-1008

47.98 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 11372144

  • 84.55 € / Pack of 50
Estimated Shipment: 31-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Peel backing off the adhesive strip across the top and hang close to your work area
  • Back panel is longer than the front for easy opening
  • High Molecular Weight High Density (HMHD) polyethylene; steam autoclavable at 121°C (250°F)
  • Maximum weight of contents should not exceed 1.36kg (3lb)
Specifications

Specifications

Laboratory Waste Bag
0.5 gal.
Zip Tie
Single-use
General Trash
HDPE
0.025 mm
10 x 8 in. (25.4 x 20.32cm)
25.4 cm
20.32 cm
Autoclavable
2 L
White
Cleanware
Self-adhesive strip
Non-sterile
1 mil
10 in.
8 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Cleanware™ Self-Adhesive Waste Bags > Laboratory Waste Bags; 203 x 254mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.