Learn More
Bel-Art™ Cleanware™ Self-Adhesive Waste Bags
Disposable waste bags free bench or hood space when hung from walls. Bel Art™ Cleanware™ Self-Adhesive Waste Bags adhere to cabinets and other surfaces.
Brand: Bel-Art™ F13174-1012
79.94 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Peel backing off the adhesive strip across the top and hang close to your work area
- Back panel is longer than the front for easy opening
- High Molecular Weight High Density (HMHD) polyethylene; steam autoclavable at 121°C (250°F)
- Maximum weight of contents should not exceed 1.36kg (3lb)
Specifications
Laboratory Waste Bag | |
1.5 gal. | |
Zip Tie | |
Single-use | |
General Trash | |
HDPE | |
0.025 mm | |
16 x 12 in. (40.64 x 30.48cm) | |
40.64 cm | |
30.48 cm |
Autoclavable | |
6 L | |
White | |
Cleanware | |
Self-adhesive strip | |
Non-sterile | |
1 mil | |
16 in. | |
12 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.