missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Conway Diffusion Cells

For Obrink method of determining carbon dioxide, ammonia or alcohol in blood or serum by diffusion method. Bel Art™ Conway Diffusion Cells are steam autoclavable at 121°C (250°F).

Brand:  Bel-Art™ F40941-0000

Product Code. 11795074

  • 43.10 € / Pack of 3
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For Obrink method of determining carbon dioxide, ammonia or alcohol in blood or serum by diffusion method. Bel Art™ Conway Diffusion Cells are steam autoclavable at 121°C (250°F).

  • Smooth, white molded polypropylene with natural, clear polypropylene cover
  • The cover has a lifting knob and fits the annular moat of the cell to seal off the chamber
  • Steam autoclavable at 121°C (250°F)

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Conway Diffusion Cells > Diffusion Cell, O.D.: 83mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.