missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Cryo-Safe™ Microcentrifuge Tube and Cryogenic Vial Coolers

Benchtop-sized coolers for microcentrifuge tubes or cryogenic vials.

154.55 € - 280.67 €

Specifications

Material Polycarbonate
Autoclavable Autoclavable
Temperature Control No
View More Specs

Products 4
Product Code Brand Color Holds Dimensions (L x W x H) Weight (Metric) Price Quantity & Availability  
Product Code Brand Color Holds Dimensions (L x W x H) Weight (Metric) Price Quantity & Availability  
11784124
View Documents
Bel Art™
F18846-0000
Clear, Red 12 Tubes (0.5 to 1.5 mL) 5.75 x 4 x 3.75 in. 0.7 kg
172.00 €
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
 
10258143
View Documents
Bel Art™
F18846-2020
White, Yellow 32 Tubes (0.5 to 2 mL) 9.5 x 3.19 x 5.7 in. 2 kg
268.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11717974
View Documents
Bel Art™
F18846-1000
Clear, Red 12 Microcentrifuge Tubes (15 mL) 7.75 x 5.5 x 7.5 in. 1.6 kg
154.55 €
Each
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
 
11794124
View Documents
Bel Art™
F18846-2000
Gray, Red 32 Tubes (0.5 to 1.5 mL) 9.5 x 3.19 x 5.7 in. 2 kg
280.67 €
Each
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
 
Description

Description

Benchtop-sized coolers for microcentrifuge tubes or cryogenic vials.

  • Holds Twelve 0.5 or 1.5mL tube in individual wells below 0 deg.C for 2.5 Hours
  • Body of cooler filled with a nontoxic gel that does not contact the contents of the cooler
  • Excellent for safe transport of temperature sensitive tubes
  • Polycarbonate body and lid is easy to clean
  • Stackable design with rubber feet to prevent sliding
  • Mini Coolers feature molded-in side handles, Junior and Maxi Coolers have fold down wire carrying handle that helps retain lid
  • Store -15°C coolers at -15°C for 24 hours prior to use
  • Store 0°C coolers at -20°C for a few hours prior to use
  • Store -20°C coolers at -20°C for 24 hours prior to use
Specifications

Specifications

Polycarbonate
Autoclavable
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel Art™ Cryo-Safe™ Microcentrifuge Tube and Cryogenic Vial Coolers

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.